Soxhlet extractors
Cone 45/40, 2000 ml, 100 NS
WITG2.212.002
Borosilicate glass. For extraction from solids with solvents.
- Very good chemical resistance
- Very good temperature resistance
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
- Extraction cap.:2000 ml
- Socket size:100 NS
- Description:Cone 45/40
- Seq:02