Orders LinkContactUsLinkComponent
Capillary installation gauges
Capillary installation gauges
Capillary installation gauge for Agilent-style fittings (compact ferrules)
  REST21034
 :   Restek
Capillary installation gauges
  REST21034
 :  Restek
Capillary installation gauges
Capillary installation gauges
Capillary installation gauge for Agilent-style fittings (compact ferrules)
Inquire For Price
Log in for availability
1 * 1 items
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
  • Prevents crushed column ends
  • Seats graphite ferrules onto column for consistent installations
Specifications
  • Description:
    Capillary installation gauge for Agilent-style fittings (compact ferrules)
  • Seq:
    01