Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Splitless, for trace samples <2 µl, for Varian 1075/1077, 6,3, 2,0 mm, 74 mm
  568-2318
 :   Restek
Inlet liners
  568-2318
 :  Restek
Inlet liners
Inlet liners

Splitless, for trace samples <2 µl, for Varian 1075/1077, 6,3, 2,0 mm, 74 mm

 
 
   
1 * 25 items
 

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

 
  • Ext.Ø:
    6,3
  • Ø int.:
    2,0 mm
  • Description:
    Splitless, for trace samples <2 µl, for Varian 1075/1077
  • Length:
    74 mm
  • Seq:
    044