Orders LinkContactUsLinkComponent
Standard ground joints
Standard ground joints
- NS, 19/26 NS, 22 mm
Catalog # QUTW1031
Standard ground joints
Catalog # QUTW1031
Standard ground joints
Standard ground joints
- NS, 19/26 NS, 22 mm
Price
93,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving high resistance to temperature changes.
Specifications
  • Cone size:
    - NS
  • Socket size:
    19/26 NS
  • Tube ext.Ø:
    22 mm
  • Seq:
    02