Distilling heads
19/26 NS, 29/32 NS
Catalog # 201-0207
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With one NS socket and two NS cones, descending angle: 75°.
Specifications
- Cone to condenser:19/26 NS
- Cone to flask:29/32 NS