Orders LinkContactUsLinkComponent
Calorimetric standards
Calorimetric standards
Benzoic acid, 1,0 gram pellets, bottle of 100, 1
Catalog # PARR3415
Supplier:   PARR
Calorimetric standards
Catalog # PARR3415
Supplier:  PARR
CAS Number:  
Calorimetric standards
Benzoic acid, 1,0 gram pellets, bottle of 100, 1
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents
Calorific standard in powder or pellet form for standardising all oxygen bomb calorimeters.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Benzoic acid, 1,0 gram pellets, bottle of 100
Specifications