Order Entry
Export
Orders LinkContactUsLinkComponent
Heating tapes, IT-S20

Heating tapes, IT-S20

Heating tape IT-S20, 3 m length, 650 W
Catalog # ISOP443830-000
Supplier:   ISOPAD
Heating tapes, IT-S20
Catalog # ISOP443830-000
Supplier:  ISOPAD
Heating tapes, IT-S20
Heating tapes, IT-S20

Heating tape IT-S20, 3 m length, 650 W

Price
280,60 €
Log in for availability
1 * 1 items
Product Details & Documents

Isopad IT-S20 is a glass silk, preterminated, flexible heating tape with a copper-tinned outer braiding serving as a protective earth braid. The temperature depends on the power and application. This enables maximum power to be installed even in the small surfaces. Provided with class I electrical protection.

  • Maximum operating temperature: 450 °C
  • Width×Thickness: 12×7 mm (±10%)
  • Maximum power per meter 220 W/m

It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.

Specifications
  • Description:
    Heating tape IT-S20, 3 m length, 650 W
  • Seq:
    04