Order Entry
Export
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Baffle splitter, universal for the most common analyses, 6,2 mm, 1,0 mm, 92,1
Catalog # 568-2322
Supplier:   Restek
Inlet liners
Catalog # 568-2322
Supplier:  Restek
Inlet liners
Inlet liners

Baffle splitter, universal for the most common analyses, 6,2 mm, 1,0 mm, 92,1

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,2 mm
  • Ø int.:
    1,0 mm
  • Description:
    Baffle splitter, universal for the most common analyses
  • Length:
    92,1
  • Seq:
    086