Order Entry
Export
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Cyclosplitter for dirty samples, many injections before cleaning required, 6,3 mm, 4,0 mm, 78,5 mm
Catalog # REST20707
Supplier:   Restek
Inlet liners
Catalog # REST20707
Supplier:  Restek
Inlet liners
Inlet liners

Cyclosplitter for dirty samples, many injections before cleaning required, 6,3 mm, 4,0 mm, 78,5 mm

Price
266,00 €
Log in for availability
1 * 5 items
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,3 mm
  • Ø int.:
    4,0 mm
  • Description:
    Cyclosplitter for dirty samples, many injections before cleaning required
  • Length:
    78,5 mm
  • Seq:
    019