Inlet liners
Cyclo double taper, for trace, active and dirty samples >2 µl, 6,5 mm, 2,0 mm, 78,5 mm
REST20908
: Restek
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
- Ext.Ø:6,5 mm
- Ø int.:2,0 mm
- Description:Cyclo double taper, for trace, active and dirty samples >2 µl
- Length:78,5 mm
- Seq:035



