Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Splitless, for trace samples <2 µl, 6,5, 2,0 mm, 78,5 mm
Catalog # 568-2316
Supplier:   Restek
Inlet liners
Catalog # 568-2316
Supplier:  Restek
Inlet liners
Inlet liners

Splitless, for trace samples <2 µl, 6,5, 2,0 mm, 78,5 mm

Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,5
  • Ø int.:
    2,0 mm
  • Description:
    Splitless, for trace samples <2 µl
  • Length:
    78,5 mm
  • Seq:
    012