O-rings, graphite
O-rings for splitless liners, I.Ø: 6,50 mm
REST20299
: Restek
These O-rings are made of graphite. Ideal for Bruker/Varian 1177 injectors.
- Maximum operating temperature 450 °C
- Can used with split or splitless liners
- Description:O-rings for splitless liners, I.Ø: 6,50 mm
- Seq:04



