Order Entry
Export
Orders LinkContactUsLinkComponent
Holders for dissecting needles

Holders for dissecting needles

Holder for dissecting needles, 11 g, 125 mm
Catalog # 233-0200
Supplier:   USBECK Laborgeräte
Holders for dissecting needles
Catalog # 233-0200
Holders for dissecting needles
Holders for dissecting needles

Holder for dissecting needles, 11 g, 125 mm

Price
7,20 €
Log in for availability
1 * 1 items
Product Details & Documents

Needle holders made from nickel plated brass. Either with or without plastic handle.

Specifications
  • Description:
    Holder for dissecting needles, 11 g
  • Length:
    125 mm
  • Seq:
    01