Glass beads
Glass beads, solid, 10 mm
Catalog # 201-0283
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
- Ø:10 mm
- Description:Glass beads, solid
- Seq:10
Solid glass.
For packing distillation columns.