Order Entry
Export
Orders LinkContactUsLinkComponent
 

Powder funnel, short stem

120 mm, 34 mm, 32 mm
  221-1164
Powder funnel, short stem
  221-1164
Powder funnel, short stem
Powder funnel, short stem

120 mm, 34 mm, 32 mm

 
19,40 €
   
1 * 1 items
 

Soda-Kalkglas.

 
  • Funnel O-Ø:
    120 mm
  • Seq:
    06
  • Stem O-Ø:
    34 mm
  • Stem length:
    32 mm