Order Entry
Export
Orders LinkContactUsLinkComponent
 

General purpose qualitative filter papers

1003, 125 mm
Catalog # 512-6115
Supplier:   Ahlstrom
General purpose qualitative filter papers
Catalog # 512-6115
Supplier:  Ahlstrom
General purpose qualitative filter papers
General purpose qualitative filter papers

1003, 125 mm

Price
30,90 €
Log in for availability
1 * 100 items
Product Details & Documents

Ahlstrom-Munksjö’s qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers covers a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Standard qualitative filter papers are suitable for quadrant folded or gravity flow applications.

These grades contain a high level of alpha-cellulose that provides high purity (ash content below 0,06%). The inherent strength of standard grades is suitable for routine quadrant folded applications but does not allow a use in vacuum work due to low wet resistance. Grade 3hw is recommended only for general purpose filtration due to a higher ash content (<0,1%) and lower filtration properties.

Specifications
  • Grade:
    1003
  • Seq:
    69
  • Size:
    125 mm
  • Cat. No.:
    512-6115
  • Filtration speed:
    80 s (Herzberg)
  • Particle retention:
    12 - 15 µm
  • Weight:
    90 g/m²