Fumaric acid
Glass bottle for solids, 500 g
Catalog # ROTH3748.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: trans-1,2-Ethylentricarboxylic acid
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116.07 g/mol
- Merck Index:13,04308
- Boiling Pt:356 °C (1013 hPa)
- Melting Pt:287 °C
- Density:1.625 g/cm³ (20 °C)
- Flash Pt:273 °C
- EINECS:203-743-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



