Order Entry
Export
Orders LinkContactUsLinkComponent
Zinc, powder

Zinc, powder

Glass bottle for solids, 2,5 kg
Catalog # ROTH9524.1
Flammable
Environment
Supplier:   Roth Carl
Zinc, powder
Catalog # ROTH9524.1
Supplier:  Roth Carl
CAS Number:  7440-66-6
Zinc, powder

Glass bottle for solids, 2,5 kg

CAS Number:  7440-66-6
Price
104,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:7440-66-6
  • Formula:Zn
  • MDL Number:MFCD00011291
  • Molecular Weight:65.39 g/mol
  • Boiling Pt:907 °C (1013 hPa)
  • Melting Pt:419.5 °C
  • Density:7.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-175-3
  • UN:1436
  • ADR:4.3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications