Order Entry
Export
Orders LinkContactUsLinkComponent
Ion selective electrodes, potassium, Orion™

Ion selective electrodes, potassium, Orion™

Potassium Sure-Flow combination electrode, 9719BNWP, 1,0 M - 10-6 M, 2,5 - 11, 0...+40 °C, 9719BNWP, combination electrode, Waterproof BNC, 1 m cable, 13×110 mm
Catalog # 662-1578
Supplier:   Thermo Orion
Ion selective electrodes, potassium, Orion™
Catalog # 662-1578
Supplier:  Thermo Orion
Ion selective electrodes, potassium, Orion™

Potassium Sure-Flow combination electrode, 9719BNWP, 1,0 M - 10-6 M, 2,5 - 11, 0...+40 °C, 9719BNWP, combination electrode, Waterproof BNC, 1 m cable, 13×110 mm

Price
1 040,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These potassium electrodes are designed to measure potassium in aqueous solutions simply, accurately and economically.

  • Built-in Sure-Flow™ reference junction prevents electrode clogging and provides fast and stabile readings
  • Combination electrode does not require a separate reference electrode, making it convenient to use with small sample volumes

Ordering information: The half cell electrode requires a separate reference electrode which can be ordered separately.

Specifications
  • Concentration:
    1,0 M - 10-6 M
  • Connection:
    Waterproof BNC, 1 m cable
  • Ø×L:
    13×110 mm
  • Type:
    9719BNWP, combination electrode
  • Description:
    Potassium Sure-Flow combination electrode, 9719BNWP
  • Temp.:
    0...+40 °C
  • pH:
    2,5 - 11
Accessories

Product Information

  • Catalog #
    Type
    Pack type
    Availability
    Price
  • Ionic strength adjustor for potassium ion selective electrodes, 475 ml bottle
    Ionic strength adjustor for potassium ion selective electrodes, 475 ml bottle
    662-0816
    Ion selective electrode reagents
    Bottle
    Log in for availability
    1 * 475 mL - 127,00 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Optimum Results E filling solution for Potassium ISE, 5 × 60 ml bottle
    Optimum Results E filling solution for Potassium ISE, 5 × 60 ml bottle
    662-0764
    300 mL
    Log in for availability
    1 * 300 mL - 127,00 €