Ion selective electrodes, nitrate, Orion™
Nitrate plastic membrane combination electrode, 9707BNWP, 1,0 M - 7x10-6 M, 2,5 - 11, 0...+40 °C, Combination electrode, 9707BNWP, Waterproof BNC, 1 m cable, 13×110 mm
Catalog # 662-1576
Supplier: Thermo Orion
Jump to:
Product Details & Documents
These nitrate electrodes are designed for measuring nitrate in aqueous solutions quickly, simply, accurately and economically.
- Stable, drift-free potentials
- Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
- Combination electrode 9707BNWP does not require a separate reference electrode, making it convenient to use with small sample volumes
Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately.
Specifications
- Concentration:1,0 M - 7x10-6 M
- Connection:Waterproof BNC, 1 m cable
- Ø×L:13×110 mm
- Type:Combination electrode, 9707BNWP
- Description:Nitrate plastic membrane combination electrode, 9707BNWP
- Temp.:0...+40 °C
- pH:2,5 - 11
Accessories
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Nitrate standard solution
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Nitrate standard solution
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Nitrate standard solution
Product Information
- Catalog #TypePack typeAvailabilityPrice
- Ionic strength adjustor for fluoroborate, nitrate & perchlorate ion selective electrodes, 475 ml bottleIonic strength adjustor for fluoroborate, nitrate & perchlorate ion selective electrodes, 475 ml bottle662-0813Ion selective electrode reagentsBottleLog in for availability1 * 475 mL - 105,00 €
Product Information
- Catalog #TypePack typeAvailabilityPrice
- Nitrate interference suppressor solution (NISS) for nitrate ion selective electrode, for more accurate and reproducible ISE measurements, 475 ml bottleNitrate interference suppressor solution (NISS) for nitrate ion selective electrode, for more accurate and reproducible ISE measurements, 475 ml bottle662-0812Ion selective electrode reagentsBottleLog in for availability1 * 475 mL - 105,00 €



