Order Entry
Export
Orders LinkContactUsLinkComponent
Ion selective electrodes, nitrate, Orion™

Ion selective electrodes, nitrate, Orion™

Nitrate plastic membrane half-cell, 9307BNWP, 1,0 M - 7x10-6 M, 2,5 - 11, 0...+40 °C, Half-cell, 9307BNWP, Waterproof BNC, 1 m cable, 12×110 mm
Catalog # 662-1589
Supplier:   Thermo Orion
Ion selective electrodes, nitrate, Orion™
Catalog # 662-1589
Supplier:  Thermo Orion
Ion selective electrodes, nitrate, Orion™
Ion selective electrodes, nitrate, Orion™

Nitrate plastic membrane half-cell, 9307BNWP, 1,0 M - 7x10-6 M, 2,5 - 11, 0...+40 °C, Half-cell, 9307BNWP, Waterproof BNC, 1 m cable, 12×110 mm

Price
878,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These nitrate electrodes are designed for measuring nitrate in aqueous solutions quickly, simply, accurately and economically.

  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • Combination electrode 9707BNWP does not require a separate reference electrode, making it convenient to use with small sample volumes

Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately.

Specifications
  • Concentration:
    1,0 M - 7x10-6 M
  • Connection:
    Waterproof BNC, 1 m cable
  • Ø×L:
    12×110 mm
  • Type:
    Half-cell, 9307BNWP
  • Description:
    Nitrate plastic membrane half-cell, 9307BNWP
  • Temp.:
    0...+40 °C
  • pH:
    2,5 - 11
Accessories

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Nitrate standard solution
    Nitrate standard solution
    662-0801
    0,1M
    475 mL
    Log in for availability
    1 * 475 mL - 102,00 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Nitrate standard solution
    Nitrate standard solution
    662-0802
    1000 mg/l
    475 mL
    Log in for availability
    1 * 475 mL - 104,00 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Nitrate standard solution
    Nitrate standard solution
    ORIO930707
    100 mg/l
    475 mL
    Log in for availability
    1 * 475 mL - 105,00 €

Product Information

  • Catalog #
    Type
    Pack type
    Availability
    Price
  • Ionic strength adjustor for fluoroborate, nitrate & perchlorate ion selective electrodes, 475 ml bottle
    Ionic strength adjustor for fluoroborate, nitrate & perchlorate ion selective electrodes, 475 ml bottle
    662-0813
    Ion selective electrode reagents
    Bottle
    Log in for availability
    1 * 475 mL - 105,00 €

Product Information

  • Catalog #
    Type
    Pack type
    Availability
    Price
  • Nitrate interference suppressor solution (NISS) for nitrate ion selective electrode, for more accurate and reproducible ISE measurements, 475 ml bottle
    Nitrate interference suppressor solution (NISS) for nitrate ion selective electrode, for more accurate and reproducible ISE measurements, 475 ml bottle
    662-0812
    Ion selective electrode reagents
    Bottle
    Log in for availability
    1 * 475 mL - 105,00 €