Orders LinkContactUsLinkComponent
1,3-Dihydroxynaphthalene ≥98%
Glass bottle for solids, 1 g
  A17739.03
1,3-Dihydroxynaphthalene ≥98%
  A17739.03
 :  132-86-5
1,3-Dihydroxynaphthalene ≥98%
1,3-Dihydroxynaphthalene ≥98%
Glass bottle for solids, 1 g
 :  132-86-5
58,30 €
1 * 1 g
Purity: 98% CAS No.: 132-86-5 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 123-126° Beilstein Reference: 2044002 Merck Reference: 14,6396 MDL No.: MFCD00003965
 :  Naphthalene-1,3-diol, Naphthalene-1,3-diol
  •  :132-86-5
  •  :C₁₀H₈O₂
  •  :MFCD00003965
  •  :160.17 g/mol
  •  :14,6396
  •  :386 °C (1013 hPa)
  •  :123…126 °C
  •  :1.12 g/cm³ (20 °C)
  •  :Ambient
  •  :205-079-7
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g