Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Traceable® K-Type Probes

VWR® Traceable® K-Type Probes

K-type fast response probe 4028, with 1,2 m coated cable, –40…+250 °C, 15×1219 mm
Catalog # 620-2012
Supplier:   VWR Collection
VWR® Traceable® K-Type Probes
Catalog # 620-2012
Supplier:  VWR Collection
VWR® Traceable® K-Type Probes

K-type fast response probe 4028, with 1,2 m coated cable, –40…+250 °C, 15×1219 mm

Price
52,50 €
Log in for availability
1 * 1 items
Specifications
  • Range:
    –40…+250 °C
  • Description:
    K-type fast response probe 4028, with 1,2 m coated cable
  • Seq:
    06
  • Ø×L:
    15×1219 mm