Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Desktop Calculators

VWR®, Desktop Calculators

Desktop calculator HS-12 ER, 12 digit, 114×5×165 mm, 113,4 g
Catalog # 818-0066
Supplier:   VWR Collection
VWR®, Desktop Calculators
Catalog # 818-0066
Supplier:  VWR Collection
VWR®, Desktop Calculators
VWR®, Desktop Calculators

Desktop calculator HS-12 ER, 12 digit, 114×5×165 mm, 113,4 g

Price
50,50 €
Log in for availability
1 * 1 items
Product Details & Documents

Tough ABS plastic case.

  • Oversized keys
  • Large digits
  • 8- or 12-digit angled display

Arithmetic functions (add, subtract, multiply, divide), percent, three key memory, square root and floating decimal point.

With arithmetic functions (add, subtract, multiply, divide), percent, three key memory, square root, and floating decimal point.

Documents

Specifications
  • Weight:
    113,4 g
  • Description:
    Desktop calculator HS-12 ER, 12 digit
  • Seq:
    02
  • W×D×H:
    114×5×165 mm