Orders LinkContactUsLinkComponent
VWR® Safe-T-Flex, Caps for Test Tubes
VWR® Safe-T-Flex, Caps for Test Tubes
Safe-T-Flex caps, 16 mm blood collecting and culture tubes, Purple
Catalog # 548-0715
Supplier:   VWR Collection
VWR® Safe-T-Flex, Caps for Test Tubes
Catalog # 548-0715
Supplier:  VWR Collection
VWR® Safe-T-Flex, Caps for Test Tubes
VWR® Safe-T-Flex, Caps for Test Tubes
Safe-T-Flex caps, 16 mm blood collecting and culture tubes, Purple
Price
64,70 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Safe-T-Flex caps feature a unique, flexible, over-locking design, locking securely on the outside of the tube lip and can be removed with one hand. A tight seal ensures sample integrity and prevents spillage or evaporation of serum samples. This special seal protects against the aerosols of highly infectious organisms such as TB and the HTLV-III virus. They are colour coded for easy specimen identification and to help guard against cross-contamination. The top of each cap can also be numbered or labelled with indelible ink. Self-locking cap holds firmly in any position and keeps contents secure while in transit. Liquid-tight seal withstands centrifugation, agitation, refrigeration and freezing.
  • Colour coded self-locking test tube caps prevent spillage or loss of samples
  • Fit all varieties of blood collecting and disposable culture tubes
Packaging: Caps are packed in bags of 1000.
Specifications
  • For:
    16 mm blood collecting and culture tubes
  • Colour:
    Purple
  • Description:
    Safe-T-Flex caps
  • Seq:
    19