VWR® Safe-T-Flex, Caps for Test Tubes
Safe-T-Flex caps, 16 mm blood collecting and culture tubes, Yellow
Catalog # 548-0726
Supplier: VWR Collection
Jump to:
Product Details & Documents
Safe-T-Flex caps feature a unique, flexible, over-locking design, locking securely on the outside of the tube lip and can be removed with one hand. A tight seal ensures sample integrity and prevents spillage or evaporation of serum samples. This special seal protects against the aerosols of highly infectious organisms such as TB and the HTLV-III virus. They are colour coded for easy specimen identification and to help guard against cross-contamination. The top of each cap can also be numbered or labelled with indelible ink. Self-locking cap holds firmly in any position and keeps contents secure while in transit. Liquid-tight seal withstands centrifugation, agitation, refrigeration and freezing.
- Colour coded self-locking test tube caps prevent spillage or loss of samples
- Fit all varieties of blood collecting and disposable culture tubes
Packaging: Caps are packed in bags of 1000.
Documents
CoC VWR® Safe-T-Flex, Caps for Test Tubes.pdf
Conformity declaration
Specifications
- For:16 mm blood collecting and culture tubes
- Colour:Yellow
- Description:Safe-T-Flex caps
- Seq:34