Orders LinkContactUsLinkComponent
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
Pink, Cellophane wrapped box
Catalog # 630-2016
Microscope slides, SuperFrost®, Menzel Gläser
Catalog # 630-2016
Supplier:  
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
Pink, Cellophane wrapped box
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Microslide Superfrost 45° pink glass cello ISO 8037/1 1,0mm 1 * 50 items Catalog #MENZAB00008232E01
Product MENZAB00008232E01 is similar to 630-2016 but manufacturer and specifications may vary.
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
  • Labelling area resistant to all common solvents
  • Six colours for identification and colour coding
  • Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
 : ISO 8037/I.
 : In cardboard boxes of 50.
Specifications
  • Packed:
    Cellophane wrapped box
  • Colour:
    Pink
  • Seq:
    210