Orders LinkContactUsLinkComponent
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
White, Cellophane wrapped box
Catalog # 630-2014
Microscope slides, SuperFrost®, Menzel Gläser
Catalog # 630-2014
Supplier:  
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
White, Cellophane wrapped box
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Microslide Superfrost 45° white glass cello ISO 8037/1 1,0mm 1 * 50 items Catalog #630-3490
Product 630-3490 is a replacement for 630-2014 and has the same packaging and specifications, but may be a different quantity.
Microslide Superfrost 45° white glass with box cello ISO 8037/1 1,0mm 1 * 2.000 items Catalog #MENZAB00008032E042
Product MENZAB00008032E042 is a replacement for 630-2014 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
  • Labelling area resistant to all common solvents
  • Six colours for identification and colour coding
  • Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
Certifications: ISO 8037/I.
Packaging: In cardboard boxes of 50.
Specifications
  • Packed:
    Cellophane wrapped box
  • Colour:
    White
  • Seq:
    202