Microscope slides, SuperFrost®, Menzel Gläser
Pink, Cellophane wrapped box
Catalog # 630-1989
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SLIDE COLOR FROSTED CUTTED PINK 76X26MM 1 * 50 items Catalog #631-1556
Product 631-1556 is similar to 630-1989 but manufacturer and specifications may vary.
Microslide superfrost cut pink glass cello ISO 8037/1 1,0mm 1 * 50 items Catalog #MENZAA00008232E011
Product MENZAA00008232E011 is similar to 630-1989 but manufacturer and specifications may vary.
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
- Labelling area resistant to all common solvents
- Six colours for identification and colour coding
- Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
Certifications: ISO 8037/I.
Packaging: In cardboard boxes of 50.
Documents
Specifications
- Packed:Cellophane wrapped box
- Colour:Pink
- Seq:14