Pipette tips, universal, Axygen®
Standard, 0,5 - 20 µl, +, Transparent, 10 racks, 96 each
Supplier: Corning
Jump to:
Product Details & Documents
These universal pipette tips are compatible with most micropipettes.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
: Available loose or in racks, sterile or non sterile versions.
Specifications
- Packed:10 racks, 96 each
- Sterile:+
- Volume:0,5 - 20 µl
- Seq:11
- Tip style:Standard
- Tip colour:Transparent