Orders LinkContactUsLinkComponent
Pipette tips, universal, Axygen®
Bevelled, wide bore (1,5 mm), 1 - 200 µl, -, Transparent, Bulk
Catalog # 613-1293
Supplier:   Corning
Pipette tips, universal, Axygen®
Catalog # 613-1293
Supplier:  Corning
Pipette tips, universal, Axygen®
Pipette tips, universal, Axygen®
Bevelled, wide bore (1,5 mm), 1 - 200 µl, -, Transparent, Bulk
Price
32,20 €
Log in for availability
1 * 1.000 items
Product Details & Documents
These universal pipette tips are compatible with most micropipettes.
  • PP, autoclavable
  • Smooth finish for minimum retention of collected sample
  • Certified free of RNase, DNase and endotoxins
  • Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
 : Available loose or in racks, sterile or non sterile versions.
Specifications
  • Packed:
    Bulk
  • Sterile:
    -
  • Volume:
    1 - 200 µl
  • Seq:
    23
  • Tip style:
    Bevelled, wide bore (1,5 mm)
  • Tip colour:
    Transparent