Pipette tips, universal, Axygen®
Bevelled, 1 - 200 µl, -, Transparent, 10 racks, 96 each
613-0650
: Corning
These universal pipette tips are compatible with most micropipettes.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
Ordering information: Available loose or in racks, sterile or non sterile versions.
- Packed:10 racks, 96 each
- Sterile:-
- Volume:1 - 200 µl
- Seq:14
- Tip style:Bevelled
- Tip colour:Transparent