Orders LinkContactUsLinkComponent
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, 0,2 ml, Violet
  731-4004
   Low Manufacturing Impact
 :   Corning
PCR microtubes with flat cap, Axygen®
  731-4004
 :  Corning
   Low Manufacturing Impact
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, 0,2 ml, Violet
70,40 €
Log in for availability
1 * 1.000 items
Polypropylene tubes are specifically designed for PCR and thermal transfer applications.
  • Made from highly polished molds
  • Free of lubricants, dyes, heavy metals, and fillers
  • Certified RNase-, DNase-, and pyrogen-free to ensure the purity of your reaction
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling.
Specifications
  • Colour:
    Violet
  • Description:
    PCR microtubes with flat cap, 0,2 ml
  • Seq:
    04
  • Cat. No.:
    731-4004
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Low Manufacturing Impact