Pipette tips, universal, Axygen®
Bevelled, Matrix-style, 15 - 1250 µl, +, Transparent, 12 racks, 96 each
Catalog # 613-1269
Supplier: Corning
Jump to:
Product Details & Documents
These universal pipette tips are compatible with most micropipettes.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
Ordering information: Available loose or in racks, sterile or non sterile versions.
Specifications
- Packed:12 racks, 96 each
- Sterile:+
- Volume:15 - 1250 µl
- Seq:70
- Tip style:Bevelled, Matrix-style
- Tip colour:Transparent