Order Entry
Export
Orders LinkContactUsLinkComponent
Pronase E

Pronase E

Plastic bottle for solids, 1 g
Catalog # ROCH10165921001
Supplier:   ROCHE
Pronase E
Catalog # ROCH10165921001
Supplier:  ROCHE
CAS Number:  
Pronase E

Plastic bottle for solids, 1 g

CAS Number:  
Price
173,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 g
  • Cat. No.:
    ROCH10165921001
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD02253017
  • Enzyme name:
    Pronase E
  • CAS No.:
    9036-06-0

Specifications