Orders LinkContactUsLinkComponent
Containers, with screw cap, LaboPlast®
125 ml, 52 mm, 67 mm
  BURK6214-0125
 :   BURKLE
Containers, with screw cap, LaboPlast®
  BURK6214-0125
 :  BURKLE
Containers, with screw cap, LaboPlast®
Containers, with screw cap, LaboPlast®
125 ml, 52 mm, 67 mm
1,00 €
1 * 1 items
PP, white.
  • Sterilisable
  • Material impermeable to light
  • Suitable for foodstguffs
Shapely packing container with screw cap. Emptying without residues because there are no crevices.
  • Ext.Ø:
    52 mm
  • Capacity:
    125 ml
  • Height:
    67 mm