Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR® Cooling Bricks

Gel pack, 165×140×25 mm, 454 g
  216-0194
VWR® Cooling Bricks
  216-0194
VWR® Cooling Bricks
VWR® Cooling Bricks

Gel pack, 165×140×25 mm, 454 g

 
45,00 €
   
1 * 36 items
 

VWR® cooling bricks are available in foam brick or gel filled pouch formats. The semi-rigid foam brick will holds its shape when refrigerated or frozen. The gel filled pouch can be refrigerated or frozen to keep products cold or frozen.

  • Long lasting, high-performance temperature protection
  • Suitable for pharmaceuticals, clinical trials, diagnostic kits, chemicals and biologicals
  • Available in a range of sizes

These packs are designed to safeguard temperature-sensitive products during storage and transport. They can be chilled in a freezer, refrigerator, or low temperature incubator and are effective when used with insulated shipping containers. Each cooling pack is vacuum sealed for consistent weight and shape. Phase changes at 0 °C and returns to foam state after thawing.

 
  • Description:
    Gel pack
  • Seq:
    07
  • W×D×H:
    165×140×25 mm
  • Weight:
    454 g