Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Test Tube Racks

VWR®, Test Tube Racks

4 (1×4), 30 mm, 180×60×80 mm, 80 mm
Catalog # 525-0195
Supplier:   VWR Collection
VWR®, Test Tube Racks
Catalog # 525-0195
Supplier:  VWR Collection
VWR®, Test Tube Racks
VWR®, Test Tube Racks

4 (1×4), 30 mm, 180×60×80 mm, 80 mm

Price
120,00 €
Log in for availability
1 * 1 items
Product Details & Documents

PTFE.

  • Excellent chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Non stick surfaces for easy cleaning

Robust and inert to virtually all chemical reagents, with very good thermal resistance. Suitable for use in cryogenics, freezers and ovens.

Specifications
  • For tube Ø:
    30 mm
  • No. of Holes:
    4 (1×4)
  • Height:
    80 mm
  • W×D×H:
    180×60×80 mm