Orders LinkContactUsLinkComponent
VWR®, Permanent Marker Pens
VWR®, Permanent Marker Pens
Marker pens, Assorted
Catalog # 811-0055
Supplier:   VWR Collection
VWR®, Permanent Marker Pens
Catalog # 811-0055
Supplier:  VWR Collection
VWR®, Permanent Marker Pens
VWR®, Permanent Marker Pens
Marker pens, Assorted
Price
58,50 €
Log in for availability
1 * 12 items
Product Details & Documents
Perfect for slides, sample bags and glassware.
  • Write on glass, plastic, porcelain, film, rubber, wood and paper
  • Permanent ink dries in seconds
Assorted colours: 6 black, 2 red, 2 blue, 2 green.
Documents
Specifications
  • Colour:
    Assorted
  • Description:
    Marker pens
  • Seq:
    07