VWR®, Spreaders, T-shaped
Spreaders, T-shaped, Blue, 1/peel pouch
Supplier: VWR Collection
Jump to:
Product Details & Documents
PS, sterile. Designed for spreading and dispersing liquids across the surface of agar culture plates. Used for performing bacterial (for example, CFU) plate counts on water, milk and other liquid samples.
- Smooth, rounded surfaces to prevent cutting or gouging of agar
- T-shaped to ensure even pressure is applied while spreading
Documents
CoC VWR Spreaders, T-Shaped.pdf
Conformity declaration
CoC_Spreaders, T-shaped, Sterile CoC_VWRI612-2651_1824942 CoC_VWRI612-2651_1920198 COC_VWRI612-2651_1922394 CoC_VWRI612-2651_2014269
More
TDS_VWR_SPREADERS_T_SHAPED
Product specification
Specifications
- Packed:1/peel pouch
- Colour:Blue
- Description:Spreaders, T-shaped