Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
PH METER, MODEL 3520, EU PLUG 1 * 1 items
PH METER, MODEL 3520, EU PLUG 1 * 1 items
Print
PH METER, MODEL 3520, EU PLUG 1 * 1 items
Catalog # JENW35459-12
Supplier:
Cole-Parmer
PH METER, MODEL 3520, EU PLUG 1 * 1 items
Catalog # JENW35459-12
Supplier:
Cole-Parmer
Expand image
Return to Top
PH METER, MODEL 3520, EU PLUG 1 * 1 items
Add to cart
Price
1 310,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Return Policy
Product Details & Documents