VWR®, Labels, Cryo
Labels for 1,5 - 2 ml tubes, on roll, Yellow, 33×13 mm, On roll
817-5008
The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage, and can also be used in most incubators.
- No need to laminate
- Use on tubes, racks and boxes
- Colour:Yellow
- Description:Labels for 1,5 - 2 ml tubes, on roll
- Dimensions:33×13 mm
- Packed:On roll



