Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Labels, Cryo

VWR®, Labels, Cryo

Labels for 1,5 - 2 ml tubes, on roll, White, 33×13 mm, On roll
Catalog # 817-5001
Supplier:   VWR Collection
VWR®, Labels, Cryo
Catalog # 817-5001
Supplier:  VWR Collection
VWR®, Labels, Cryo
VWR®, Labels, Cryo

Labels for 1,5 - 2 ml tubes, on roll, White, 33×13 mm, On roll

Price
70,40 €
Log in for availability
1 * 1.000 items
Product Details & Documents

The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage, and can also be used in most incubators.

  • No need to laminate
  • Use on tubes, racks and boxes

Documents

Specifications
  • Colour:
    White
  • Description:
    Labels for 1,5 - 2 ml tubes, on roll
  • Dimensions:
    33×13 mm
  • Packed:
    On roll