Electrodes, Genetrodes™
Genetrode™ kit, straight, 10 mm with holder and cables
BTXI45-0161
: BTX
Genetrodes™ are paired, reusable, needle style electrodes that are ideal for in vivo and in ovo electroporation applications, including drug and gene delivery.
- Five models to suit the size and shape of the target electroporation area
- Each model consists of a pair of electrodes configured as either straight or bent L-shaped electrodes with gold tips
- The electrodes are placed into tissue following injection of the molecule of interest, and an electroporation pulse is delivered using an electroporation generator
- The electric field introduced by the Genetrodes™ causes transient pores to form in the cells of the tissue, allowing uptake of the molecules into cells
- Genetrodes™ are positioned in parallel at a predetermined gap in tissue using the Genetrode/Genepaddle holder
Generator compatibility: ECM® 830, ECM® 2001
Voltage range: 0 to 200 V DC
Pulse length range: 10 µsec to 99 msec in PBS
Pulse number range: 1 to 99 (depending on voltage and pulse length settings)
Genetrode™ holder electrode gap: 1 to 10 mm range.
* Requires Genetrodes/Genepaddle holder, square post cables and coaxial banana plug cable.
Genetrode™ holder electrode gap: 1 - 10 mm range
- Application:in vivo
- Description:Genetrode™ kit, straight, 10 mm with holder and cables
- Seq:10



