Order Entry
Export
Orders LinkContactUsLinkComponent
Affinity chromatography media, MabSelect Xtra™

Affinity chromatography media, MabSelect Xtra™

MabSelect Xtra™, Liquid, 75 µm, 25 ml
Catalog # 17-5269-07
Supplier:   Cytiva
Affinity chromatography media, MabSelect Xtra™
Catalog # 17-5269-07
Supplier:  Cytiva
CAS Number:  
Affinity chromatography media, MabSelect Xtra™
Affinity chromatography media, MabSelect Xtra™

MabSelect Xtra™, Liquid, 75 µm, 25 ml

CAS Number:  
Price
3 350,00 €
Log in for availability
1 * 25 mL
Product Details & Documents

MabSelect Xtra has the same recombinant Protein A ligand as MabSelect but is based on a high-flow agarose base matrix with increased porosity and a slightly decreased particle size compared to MabSelect.

  • Higher dynamic binding capacity than currently available Protein A chromatography media
  • High purity capture due to high ligand selectivity and matrix hydrophilicity
  • Epoxy activation coupling method
  • Slightly lower pressure/flow properties compared to MabSelect SuRe and MabSelect
  • Improved process economics through reduced raw materials costs and/or reduced number of cycles
  • Prepacked, ready-to-use columns for convenient process development
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption

MabSelect Xtra is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, MabSelect Xtra meets industrial demands with security of supply and comprehensive technical and regulatory support. The medium provides a lower overall production cost due to the possibility of processing concentrated feedstocks in fewer batches.

This results in higher capacity—MabSelect Xtra has about 30% higher dynamic binding capacity for IgG than currently available Protein A chromatography media. In addition, MabSelect Xtra has a higher capacitiy for many Fc-fusion proteins.

Specifications

Specifications

  • Particle Size:
    75 µm
  • Pk:
    25 ml
  • Description:
    MabSelect Xtra™
  • Type:
    Liquid
  • Cat. No.:
    17-5269-07
  • Matrix:
    Highly cross-linked agarose
  • Binding capacity:
    40 mg human lgG/ml
  • Flow rate:
    <300 cm/h
  • Ligand:
    Recombinant protein A (E.coli)
  • pH range:
    3 - 10
  • Storage temperature:
    2 - 8 °C

Specifications