ContactUsLinkComponent
Thermometer, precision, mercury filled, DIN certified, measuring range:-50:150/0.1C, enclosed scale, capllary lens front, colourle s s, Richter-top finish with button, dimension:550x9-9.5mm, suitable for government verification. Until 10th October 1 * 1 items
Catalog # AMARL26012
Supplier:  Amarell
Thermometer, precision, mercury filled, DIN certified, measuring range:-50:150/0.1C, enclosed scale, capllary lens front, colourle s s, Richter-top finish with button, dimension:550x9-9.5mm, suitable for government verification. Until 10th October 1 * 1 items
Catalog # AMARL26012
Supplier:  Amarell