Ficoll® PM400
Ficoll™ PM400 (dry powder)
Supplier: Cytiva
Jump to:
Product Details & Documents
Ficoll® PM400 is a synthetic neutral, highly branched hydrophilic polymer of sucrose with an average molecular weight of 400000. It is used to form density gradients for separating and isolating eukaryotic cells, organelles and bacterial cells, as a stabilising agent and as a preparation medium for isolating mononuclear cells. It is useful for separating cells that are sensitive to centrifugation. Ficoll™ PM400 can also be used in other applications, such in defined culture media, nucleic acid hybridisation, electrophoresis, and immunological studies.
- Concentrations of up to 50% (w/v) covering densities of up to 1,2 g/ml can be achieved
- Better osmotic properties than sucrose
- Preserves functional and morphological integrity
- Does not penetrate biological membranes
- Serves as the raw material for preparation of Ficoll-Paque gradients
Documents
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Description:Ficoll™ PM400 (dry powder)
- Seq:02
- Cat. No.:17-0300-10
- Stability:Stable in alkaline and neutral solutions, but are rapidly hydrolyzed in solution at pH 3, especially at elevated temperature



