Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Matrix metalloproteinase 9 (MMP9) polycl 1 * 100 mg
Catalog # ORBGORB-PAB-10194
Supplier:
ORBIGEN
Expand image
Matrix metalloproteinase 9 (MMP9) polycl 1 * 100 mg
Catalog # ORBGORB-PAB-10194
Supplier:
ORBIGEN