Order Entry
Export
Orders LinkContactUsLinkComponent
4E-BP1 (PHAS-I) 1 ml 1 * 1 mL

4E-BP1 (PHAS-I) 1 ml 1 * 1 mL

Catalog # ORBGORB-BVC-10434
Supplier:   ORBIGEN
4E-BP1 (PHAS-I) 1 ml 1 * 1 mL
Catalog # ORBGORB-BVC-10434
Supplier:  ORBIGEN
4E-BP1 (PHAS-I) 1 ml 1 * 1 mL
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents