Order Entry
Export
Orders LinkContactUsLinkComponent
U2AF35 1 ml 1 * 1 mL

U2AF35 1 ml 1 * 1 mL

Catalog # ORBGORB-BVC-10284
Supplier:   ORBIGEN
U2AF35 1 ml 1 * 1 mL
Catalog # ORBGORB-BVC-10284
Supplier:  ORBIGEN
U2AF35 1 ml 1 * 1 mL
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents